Assay Details
| Gene Symbol: ADRB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_428541.6 | XP_428541.4 | 2 - 3 | 1458 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADRB3| Gene Symbol: ADRB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_428541.6 | XP_428541.4 | 2 - 3 | 1458 | 92 | |
Set Membership: |
|