Assay Details
| Gene Symbol: DUOXA1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943614.3 | XP_004943671.1 | 6 - 7 | 946 | 93 | |
| XM_015279086.2 | XP_015134572.1 | 6 - 7 | 955 | 93 | ||
| XM_025153964.1 | XP_025009732.1 | 6 - 7 | 792 | 93 | ||
More Information
Set Membership: |
|

DUOXA1L
Genomic Map