Assay Details
| Gene Symbol: FAM19A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234785.5 | XP_001234786.3 | 1 - 2 | 3392 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001234785.5 transcripts.
Set Membership: |
|

FAM19A4
Genomic Map