Assay Details
| Gene Symbol: MYO15L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015295329.2 | XP_015150815.1 | 13 - 14 | 1618 | 156 | |
| XM_015295330.2 | XP_015150816.1 | 13 - 14 | 1576 | 156 | ||
| XM_025141801.1 | XP_024997569.1 | 11 - 12 | 1922 | 156 | ||
| XM_025141802.1 | XP_024997570.1 | 11 - 12 | 1919 | 156 | ||
More Information
Set Membership: |
|

MYO15L
Genomic Map