Assay Details
| Gene Symbol: ANKRD34B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015280667.2 | XP_015136153.1 | 2 - 3 | 5178 | 113 | |
| XM_025144952.1 | XP_025000720.1 | 2 - 3 | 5178 | 113 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015280667.2, XM_025144952.1 transcripts.
Set Membership: |
|

ANKRD34B
Genomic Map