Assay Details
| Gene Symbol: POU4F3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294056.2 | XP_015149542.2 | 1 - 2 | 631 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POU4F3| Gene Symbol: POU4F3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294056.2 | XP_015149542.2 | 1 - 2 | 631 | 70 | |
Set Membership: |
|