Assay Details
| Gene Symbol: LOC107050721 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145990.1 | XP_025001758.1 | 11 - 12 | 1746 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107050721| Gene Symbol: LOC107050721 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145990.1 | XP_025001758.1 | 11 - 12 | 1746 | 69 | |
Set Membership: |
|