Assay Details
| Gene Symbol: SH3RF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945041.3 | XP_004945098.1 | 3 - 4 | 898 | 59 | |
| XM_015294060.2 | XP_015149546.1 | 2 - 3 | 1139 | 59 | ||
| XM_414662.6 | XP_414662.3 | 2 - 3 | 1138 | 59 | ||
More Information
Set Membership: |
|

SH3RF2
Genomic Map