Assay Details
| Gene Symbol: COL2A1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015279683.2 | XP_015135169.1 | 21 - 22 | 2169 | 164 | |
| XM_025156090.1 | XP_025011858.1 | 21 - 22 | 2169 | 164 | ||
| XM_025156091.1 | XP_025011859.1 | 21 - 22 | 2169 | 164 | ||
| XM_025156092.1 | XP_025011860.1 | 21 - 22 | 2169 | 164 | ||
| XM_025156093.1 | XP_025011861.1 | 21 - 22 | 2169 | 164 | ||
| XM_025156094.1 | XP_025011862.1 | 21 - 22 | 2169 | 164 | ||
| XM_025156095.1 | XP_025011863.1 | 21 - 22 | 2169 | 164 | ||
| XM_025156096.1 | XP_025011864.1 | 21 - 22 | 2375 | 164 | ||
| XM_025156097.1 | XP_025011865.1 | 17 - 18 | 2026 | 164 | ||
More Information
Set Membership: |
|

COL2A1L
Genomic Map