Assay Details
| Gene Symbol: PRDM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025149228.1 | XP_025004996.1 | 2 - 3 | 1523 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRDM13| Gene Symbol: PRDM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025149228.1 | XP_025004996.1 | 2 - 3 | 1523 | 65 | |
Set Membership: |
|