Assay Details
| Gene Symbol: WIPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004939386.3 | XP_004939443.1 | 7 - 8 | 1651 | 86 | |
| XM_015281822.2 | XP_015137308.1 | 7 - 8 | 1516 | 86 | ||
| XM_025147453.1 | XP_025003221.1 | 7 - 8 | 1513 | 86 | ||
| XM_025147454.1 | XP_025003222.1 | 8 - 9 | 1793 | 86 | ||
| XM_418733.6 | XP_418733.5 | 5 - 6 | 2484 | 86 | ||
More Information
Set Membership: |
|

WIPF3
Genomic Map