Assay Details
| Gene Symbol: P3H4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_427657.6 | XP_427657.5 | 6 - 7 | 1414 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

P3H4| Gene Symbol: P3H4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_427657.6 | XP_427657.5 | 6 - 7 | 1414 | 62 | |
Set Membership: |
|