Assay Details
| Gene Symbol: FAM83G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945207.3 | XP_004945264.1 | 2 - 3 | 803 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM83G| Gene Symbol: FAM83G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945207.3 | XP_004945264.1 | 2 - 3 | 803 | 83 | |
Set Membership: |
|