Assay Details
| Gene Symbol: LOC101750148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145064.1 | XP_025000832.1 | 1 - 2 | 116 | 92 | |
| XM_025145065.1 | XP_025000833.1 | 1 - 2 | 116 | 92 | ||
More Information
Set Membership: |
|

LOC101750148
Genomic Map