Assay Details
| Gene Symbol: CCDC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015275577.2 | XP_015131063.1 | 5 - 6 | 1372 | 68 | |
| XM_415165.6 | XP_415165.2 | 5 - 6 | 1374 | 68 | ||
More Information
Set Membership: |
|

CCDC63
Genomic Map