Assay Details
| Gene Symbol: FAM155B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278333.2 | XP_015133819.1 | 1 - 2 | 897 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM155B| Gene Symbol: FAM155B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278333.2 | XP_015133819.1 | 1 - 2 | 897 | 86 | |
Set Membership: |
|