Assay Details
Gene Symbol: LOC426897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015300378.2 | XP_015155864.2 | 5 - 6 | 1433 | 64 | |
XM_424505.6 | XP_424505.5 | 5 - 6 | 1428 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
