Assay Details
| Gene Symbol: LOC426897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300378.2 | XP_015155864.2 | 7 - 8 | 1781 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC426897| Gene Symbol: LOC426897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300378.2 | XP_015155864.2 | 7 - 8 | 1781 | 99 | |
Set Membership: |
|