Assay Details
| Gene Symbol: MFRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642601.3 | XP_003642649.2 | 7 - 8 | 1143 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MFRP| Gene Symbol: MFRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642601.3 | XP_003642649.2 | 7 - 8 | 1143 | 54 | |
Set Membership: |
|