Assay Details
| Gene Symbol: ARX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025146479.1 | XP_025002247.1 | 4 - 5 | 1388 | 96 | |
| XM_025146483.1 | XP_025002251.1 | 5 - 6 | 1305 | 96 | ||
More Information
Set Membership: |
|

ARX
Genomic Map