Assay Details
| Gene Symbol: GABRA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936058.3 | XP_004936115.1 | 2 - 3 | 370 | 95 | |
| XM_420724.6 | XP_420724.2 | 2 - 3 | 365 | 95 | ||
More Information
Set Membership: |
|

GABRA4
Genomic Map