Assay Details
| Gene Symbol: TMEM45A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298111.2 | XP_015153597.1 | 3 - 4 | 519 | 58 | |
| XM_015298112.1 | XP_015153598.1 | 3 - 4 | 598 | 58 | ||
| XM_417866.6 | XP_417866.1 | 3 - 4 | 519 | 58 | ||
More Information
Set Membership: |
|

TMEM45A
Genomic Map