Assay Details
| Gene Symbol: LOC101749593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025150176.1 | XP_025005944.1 | 2 - 3 | 4361 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101749593| Gene Symbol: LOC101749593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025150176.1 | XP_025005944.1 | 2 - 3 | 4361 | 116 | |
Set Membership: |
|