Assay Details
| Gene Symbol: MIOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015281706.2 | XP_015137192.1 | 9 - 10 | 901 | 86 | |
| XM_015281753.2 | XP_015137239.1 | 9 - 10 | 926 | 86 | ||
More Information
Set Membership: |
|

MIOX
Genomic Map