Assay Details
| Gene Symbol: MICA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155879.1 | XP_025011647.1 | 5 - 6 | 991 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MICA| Gene Symbol: MICA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155879.1 | XP_025011647.1 | 5 - 6 | 991 | 90 | |
Set Membership: |
|