Assay Details
| Gene Symbol: GPR55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015277072.2 | XP_015132558.1 | 2 | 3312 | 71 | |
| XM_025153423.1 | XP_025009191.1 | 3 | 2469 | 71 | ||
More Information
Set Membership: |
|

GPR55
Genomic Map