Assay Details
| Gene Symbol: PDIL-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251099.1 | NP_001238028.1 | 8 - 9 | 1324 | 101 | |
| GenBank mRNA | AB185851.1 | - | 8 - 9 | 1324 | 101 | |
More Information
Set Membership: |
|

PDIL-2
Genomic Map