Assay Details
| Gene Symbol: HDT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248955.1 | NP_001235884.1 | 8 - 9 | 842 | 88 | |
| GenBank mRNA | AF532618.1 | - | 8 - 9 | 842 | 88 | |
More Information
Set Membership: |
|

HDT1
Genomic Map