Assay Details
| Gene Symbol: PDS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249840.1 | NP_001236769.1 | 12 - 13 | 1871 | 70 | |
| GenBank mRNA | M64704.1 | - | 12 - 13 | 1871 | 70 | |
| Gene Symbol: LOC100127420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK244738.1 | - | 13 - 14 | 1763 | 70 | |
More Information
Set Membership: |
|

PDS1
Genomic Map