Assay Details
| Gene Symbol: AAPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251355.1 | NP_001238284.1 | 16 - 17 | 1181 | 96 | |
| GenBank mRNA | AK246054.1 | - | 16 - 17 | 1136 | 96 | |
| U12735.1 | - | 16 - 17 | 1181 | 96 | ||
More Information
Set Membership: |
|

AAPT1
Genomic Map