Assay Details
| Gene Symbol: LOC547869 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248454.1 | NP_001235383.1 | 2 - 3 | 515 | 70 | |
| GenBank mRNA | U50081.1 | - | 2 - 3 | 515 | 70 | |
| X13302.1 | - | 2 - 3 | 503 | 70 | ||
More Information
Set Membership: |
|

LOC547869
Genomic Map