Assay Details
| Gene Symbol: USP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250505.1 | NP_001237434.1 | 4 - 5 | 528 | 73 | |
| GenBank mRNA | DQ267699.1 | - | 4 - 5 | 528 | 73 | |
More Information
Set Membership: |
|

USP1
Genomic Map