Assay Details
| Gene Symbol: OAS-TL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251463.1 | NP_001238392.1 | 3 - 4 | 276 | 80 | |
| GenBank mRNA | AK243703.1 | - | 3 - 4 | 196 | 80 | |
| AK285461.1 | - | 3 - 4 | 189 | 80 | ||
| BT099087.1 | - | 8 - 9 | 80 | |||
| EF433420.1 | - | 3 - 4 | 276 | 80 | ||
More Information
Set Membership: |
|

OAS-TL1
Genomic Map