Assay Details
| Gene Symbol: LOX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250394.1 | NP_001237323.1 | 6 - 7 | 1358 | 97 | |
| GenBank mRNA | AK243941.1 | - | 6 - 7 | 1358 | 97 | |
| AK244195.1 | - | 6 - 7 | 1359 | 97 | ||
| AK244303.1 | - | 6 - 7 | 1362 | 97 | ||
| EU003576.1 | - | 6 - 7 | 1358 | 97 | ||
More Information
Set Membership: |
|

LOX9
Genomic Map