Assay Details
| Gene Symbol: WNK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250545.1 | NP_001237474.1 | 3 - 4 | 554 | 78 | |
| GenBank mRNA | FJ795035.1 | - | 3 - 4 | 554 | 78 | |
More Information
Set Membership: |
|

WNK5
Genomic Map