Assay Details
| Gene Symbol: BZIP28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250309.1 | NP_001237238.1 | 948 | 104 | ||
| GenBank mRNA | DQ787065.1 | - | 948 | 104 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BZIP28| Gene Symbol: BZIP28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250309.1 | NP_001237238.1 | 948 | 104 | ||
| GenBank mRNA | DQ787065.1 | - | 948 | 104 | ||
Set Membership: |
|