Assay Details
| Gene Symbol: LOC100812152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253930.1 | NP_001240859.1 | 5 - 6 | 415 | 113 | |
| GenBank mRNA | BT095167.1 | - | 5 - 6 | 747 | 113 | |
More Information
Set Membership: |
|

LOC100812152
Genomic Map