Assay Details
| Gene Symbol: LOC100812152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253930.1 | NP_001240859.1 | 6 - 7 | 546 | 112 | |
| GenBank mRNA | BT095167.1 | - | 4 - 5 | 616 | 112 | |
More Information
Set Membership: |
|

LOC100812152
Genomic Map