Assay Details
| Gene Symbol: LOC100804033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254369.1 | NP_001241298.1 | 1 - 2 | 208 | 101 | |
| GenBank mRNA | BT096465.1 | - | 1 - 2 | 208 | 101 | |
More Information
Set Membership: |
|

LOC100804033
Genomic Map