Assay Details
| Gene Symbol: LOC100803033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254498.2 | NP_001241427.1 | 1 - 2 | 56 | 70 | |
| GenBank mRNA | AK245714.1 | - | 1 - 2 | 57 | 70 | |
| BT097805.1 | - | 1 - 2 | 58 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT097805.1, NM_001254498.2 transcripts.
Set Membership: |
|

LOC100803033
Genomic Map