Assay Details
| Gene Symbol: LOC100782621 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255064.2 | NP_001241993.1 | 3 - 4 | 338 | 113 | |
| GenBank mRNA | BT098798.1 | - | 3 - 4 | 340 | 113 | |
More Information
Set Membership: |
|

LOC100782621
Genomic Map