Assay Details
| Gene Symbol: LOC100801138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255405.1 | NP_001242334.1 | 1 - 2 | 204 | 97 | |
| GenBank mRNA | BT097829.1 | - | 1 - 2 | 204 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT097829.1, NM_001255405.1 transcripts.
Set Membership: |
|

LOC100801138
Genomic Map