Assay Details
| Gene Symbol: LOC100801138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255405.1 | NP_001242334.1 | 6 - 7 | 766 | 143 | |
| GenBank mRNA | BT097829.1 | - | 6 - 7 | 766 | 143 | |
More Information
Set Membership: |
|

LOC100801138
Genomic Map