Assay Details
| Gene Symbol: LOC100787109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255616.2 | NP_001242545.1 | 1 - 2 | 302 | 97 | |
| GenBank mRNA | BT096213.1 | - | 1 - 2 | 304 | 97 | |
| Gene Symbol: LOC100798544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK244359.1 | - | 1 - 2 | 340 | 97 | |
More Information
Set Membership: |
|

LOC100787109
Genomic Map