Assay Details
| Gene Symbol: LOC100792937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253270.2 | NP_001240199.1 | 1 - 2 | 173 | 58 | |
| GenBank mRNA | BT097485.1 | - | 1 - 2 | 175 | 58 | |
More Information
Set Membership: |
|

LOC100792937
Genomic Map