Assay Details
| Gene Symbol: LOC100795506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255404.2 | NP_001242333.1 | 1 - 2 | 71 | 94 | |
| GenBank mRNA | BT097676.1 | - | 1 - 2 | 73 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT097676.1, NM_001255404.2 transcripts.
Set Membership: |
|

LOC100795506
Genomic Map