Assay Details
| Gene Symbol: LOC100819709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255960.2 | NP_001242889.1 | 6 - 7 | 741 | 93 | |
| GenBank mRNA | BT093615.1 | - | 6 - 7 | 743 | 93 | |
More Information
Set Membership: |
|

LOC100819709
Genomic Map