Assay Details
| Gene Symbol: LOC100819709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255960.2 | NP_001242889.1 | 8 - 9 | 961 | 104 | |
| GenBank mRNA | BT093615.1 | - | 8 - 9 | 963 | 104 | |
More Information
Set Membership: |
|

LOC100819709
Genomic Map