Assay Details
| Gene Symbol: LOC100170748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251765.1 | NP_001238694.1 | 2 | 217 | 84 | |
| GenBank mRNA | EU444600.1 | - | 2 | 217 | 84 | |
More Information
Set Membership: |
|

LOC100170748
Genomic Map