Assay Details
| Gene Symbol: LOC100787970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252702.1 | NP_001239631.1 | 1 | 123 | 98 | |
| GenBank mRNA | BT093792.1 | - | 1 | 123 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT093792.1, NM_001252702.1 transcripts.
Set Membership: |
|

LOC100787970
Genomic Map